missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoropiperidine hydrochloride, 95%

Catalog No. p-7028435
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH25755MD 250 mg
AAH2575503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH25755MD Supplier Thermo Scientific Chemicals Supplier No. H25755MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57395-89-8 | C5H11ClFN | 139.598 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57395-89-8
Molecular Formula C5H11ClFN
Molecular Weight (g/mol) 139.598
MDL Number MFCD03452786
InChI Key IXENWFQXVCOHAZ-UHFFFAOYSA-N
Synonym 4-fluoropiperidine hydrochloride, 4-fluoropiperidine hcl, 4-fluoropiperidinehydrochloride, acmc-1auoh, 4-fluoro-piperidine hydrochloride, 4-fluoranylpiperidine hydrochloride, piperidine, 4-fluoro-, hydrochloride, 78197-27-0 4-fluoropiperidine hydrochloride
PubChem CID 2759158
IUPAC Name 4-fluoropiperidine;hydrochloride
SMILES C1CNCCC1F.Cl

Specifications

Melting Point 161°C to 163°C
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 139.6
Percent Purity 95%
Chemical Name or Material 4-Fluoropiperidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only