Learn More
4-Fluorophthalic anhydride, 98%
CAS: 319-03-9 | C8H3FO3 | 166.11 g/mol
Supplier: Thermo Scientific Chemicals B2135214
Description
4-Fluorophthalic anhydride is used in the preparation of 5-fluoro-2-pyridin-3-ylmethyl-isoindole-1,3-dione by reacting with C-pyridin-3-yl-methylamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluorophthalic anhydride is used in the preparation of 5-fluoro-2-pyridin-3-ylmethyl-isoindole-1,3-dione by reacting with C-pyridin-3-yl-methylamine.
Solubility
Soluble in sodium hydroxide.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
| 4-Fluorophthalic anhydride | |
| 74°C to 79°C | |
| 147°C to 149°C (20 mmHg) | |
| MFCD00191363 | |
| 131281 | |
| 4-fluorophthalic anhydride, 5-fluoroisobenzofuran-1,3-dione, 5-fluoro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 5-fluoro, 4-fluorophthalicanhydride, 4-fluorophthalic acid anhydride, 5-fluoro-1,3-dihydro-2-benzofuran-1,3-dione, 4-fluorobenzene-1,2-dicarboxylic anhydride, 5-fluoro-isobenzofuran-1,3-dione | |
| XVMKZAAFVWXIII-UHFFFAOYSA-N | |
| 5-fluoro-2-benzofuran-1,3-dione | |
| 67572 | |
| 98% |
| 319-03-9 | |
| 1.5 | |
| C8H3FO3 | |
| 25 g | |
| Moisture sensitive | |
| Soluble in sodium hydroxide. | |
| FC1=CC=C2C(=O)OC(=O)C2=C1 | |
| 166.11 | |
| 166.11 |
Chemical Identifiers
| 319-03-9 | |
| 166.11 | |
| XVMKZAAFVWXIII-UHFFFAOYSA-N | |
| 67572 | |
| FC1=CC=C2C(=O)OC(=O)C2=C1 |
| C8H3FO3 | |
| MFCD00191363 | |
| 4-fluorophthalic anhydride, 5-fluoroisobenzofuran-1,3-dione, 5-fluoro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 5-fluoro, 4-fluorophthalicanhydride, 4-fluorophthalic acid anhydride, 5-fluoro-1,3-dihydro-2-benzofuran-1,3-dione, 4-fluorobenzene-1,2-dicarboxylic anhydride, 5-fluoro-isobenzofuran-1,3-dione | |
| 5-fluoro-2-benzofuran-1,3-dione |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c
H302-H315-H317-H319-H334-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only