Learn More
4-Fluorophthalic acid, 98%
CAS: 320-97-8 | C8H5FO4 | 184.12 g/mol
Supplier: Thermo Scientific Chemicals H3159709
Description
4-Fluorophthalic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluorophthalic acid is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
| 4-Fluorophthalic acid | |
| 154°C to 158°C | |
| C8H5FO4 | |
| 10 g | |
| Insoluble in water. | |
| OC(=O)C1=CC=C(F)C=C1C(O)=O | |
| 184.12 | |
| 184.12 |
| 320-97-8 | |
| Odorless | |
| MFCD06208213 | |
| 4-fluorobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-fluoro, 5-fluorophtalic acid, 5-fluorophthalic acid, 4-fluorobenzene-1,2-dioic acid, 3-fluorophthalicacid, pubchem1406, rarechem al bo 0829, 1,2-benzenedicarboxylicacid, 4-fluoro | |
| OMCXTFVBNCFZMY-UHFFFAOYSA-N | |
| 4-fluorophthalic acid | |
| 15312 | |
| 98% |
Chemical Identifiers
| 320-97-8 | |
| 184.12 | |
| OMCXTFVBNCFZMY-UHFFFAOYSA-N | |
| 15312 | |
| OC(=O)C1=CC=C(F)C=C1C(O)=O |
| C8H5FO4 | |
| MFCD06208213 | |
| 4-fluorobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-fluoro, 5-fluorophtalic acid, 5-fluorophthalic acid, 4-fluorobenzene-1,2-dioic acid, 3-fluorophthalicacid, pubchem1406, rarechem al bo 0829, 1,2-benzenedicarboxylicacid, 4-fluoro | |
| 4-fluorophthalic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only