missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorophenylhydrazine Hydrochloride 97.0+%, TCI America™
Supplier: TCI America F024625G
Specifications
| 4-Fluorophenylhydrazine Hydrochloride | |
| C6H8ClFN2 | |
| 25 g | |
| FEKUXLUOKFSMRO-UHFFFAOYSA-N | |
| hydrogen (4-fluorophenyl)hydrazine chloride | |
| 69981 | |
| ≥97.0% (HPLC,T) |
| 823-85-8 | |
| MFCD00012942 | |
| 4-fluorophenylhydrazine hydrochloride, 4-fluorophenyl hydrazine hydrochloride, 4-fluorophenylhydrazine hcl, 1-4-fluorophenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine hcl, p-fluorophenylhydrazine hydrochloride, 4-fluorphenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine monohydrochloride, p-fluorophenylhydrazine hcl, hydrazine, 4-fluorophenyl-, monohydrochloride | |
| [H+].[Cl-].NNC1=CC=C(F)C=C1 | |
| 162.59 | |
| 162.59 | |
| Crystalline Powder |
Chemical Identifiers
| 823-85-8 | |
| 162.59 | |
| FEKUXLUOKFSMRO-UHFFFAOYSA-N | |
| 69981 | |
| [H+].[Cl-].NNC1=CC=C(F)C=C1 |
| C6H8ClFN2 | |
| MFCD00012942 | |
| 4-fluorophenylhydrazine hydrochloride, 4-fluorophenyl hydrazine hydrochloride, 4-fluorophenylhydrazine hcl, 1-4-fluorophenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine hcl, p-fluorophenylhydrazine hydrochloride, 4-fluorphenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine monohydrochloride, p-fluorophenylhydrazine hcl, hydrazine, 4-fluorophenyl-, monohydrochloride | |
| hydrogen (4-fluorophenyl)hydrazine chloride |
Safety and Handling
TSCA : No