Learn More
4-Fluorophenylhydrazine hydrochloride, 97%
CAS: 823-85-8 | C6H8ClFN2 | 162.59 g/mol
Supplier: Thermo Scientific Chemicals A1562414
Description
4-Fluorophenylhydrazine hydrochloride is used as a chemical intermediate in the pharmaceutica1ls particularly of tryptamine drugs used in the treatment of migraine and cluster headaches. It is used to prepare agrochemical, and other chemical. It is used to detect sugars and aldehydes in analytical chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluorophenylhydrazine hydrochloride is used as a chemical intermediate in the pharmaceutica1ls particularly of tryptamine drugs used in the treatment of migraine and cluster headaches. It is used to prepare agrochemical, and other chemical. It is used to detect sugars and aldehydes in analytical chemistry.
Solubility
Soluble in water.
Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.
Specifications
| 4-Fluorophenylhydrazine hydrochloride | |
| ∼265°C (decomposition) | |
| MFCD00012942 | |
| 3627721 | |
| Soluble in water. | |
| [H+].[Cl-].NNC1=CC=C(F)C=C1 | |
| 162.59 | |
| 162.6 |
| 823-85-8 | |
| C6H8ClFN2 | |
| 25 g | |
| 4-fluorophenylhydrazine hydrochloride, 4-fluorophenyl hydrazine hydrochloride, 4-fluorophenylhydrazine hcl, 1-4-fluorophenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine hcl, p-fluorophenylhydrazine hydrochloride, 4-fluorphenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine monohydrochloride, p-fluorophenylhydrazine hcl, hydrazine, 4-fluorophenyl-, monohydrochloride | |
| FEKUXLUOKFSMRO-UHFFFAOYSA-N | |
| (4-fluorophenyl)hydrazine;hydrochloride | |
| 69981 | |
| 98% |
Chemical Identifiers
| 823-85-8 | |
| 162.59 | |
| FEKUXLUOKFSMRO-UHFFFAOYSA-N | |
| 69981 | |
| [H+].[Cl-].NNC1=CC=C(F)C=C1 |
| C6H8ClFN2 | |
| MFCD00012942 | |
| 4-fluorophenylhydrazine hydrochloride, 4-fluorophenyl hydrazine hydrochloride, 4-fluorophenylhydrazine hcl, 1-4-fluorophenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine hcl, p-fluorophenylhydrazine hydrochloride, 4-fluorphenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine monohydrochloride, p-fluorophenylhydrazine hcl, hydrazine, 4-fluorophenyl-, monohydrochloride | |
| (4-fluorophenyl)hydrazine;hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P362-P501c
H302+H312+H332-H315-H317-H319-H335-H351
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 212-521-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only