missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$69.31 - $69.31
Specifications
| CAS | 1765-93-1 |
|---|---|
| Melting Point | 265°C |
| Molecular Formula | C6H6BFO2 |
| MDL Number | MFCD00039136 |
| InChI Key | LBUNNMJLXWQQBY-UHFFFAOYSA-N |
Specifications
| 1765-93-1 | |
| C6H6BFO2 | |
| LBUNNMJLXWQQBY-UHFFFAOYSA-N | |
| 139.92 | |
| CHEBI:48661 | |
| Crystalline Powder |
| 265°C | |
| MFCD00039136 | |
| (4-fluorophenyl)boronic acid | |
| 285645 | |
| 139.92 |