missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F03615G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F03615G Supplier TCI America Supplier No. F03615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-Fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 1765-93-1
Melting Point 265°C
Molecular Formula C6H6BFO2
Quantity 5 g
MDL Number MFCD00039136
Synonym 4-fluorobenzeneboronic acid, 4-fluorophenyl boronic acid, 4-fluorophenyl boranediol, p-fluorophenylboronic acid, 4-fluorophenylbornic acid, 4-fluorophenyl dihydroxyboron, 4-fluorophenyl dihydroxyborane, p-fluorophenyl boric acid, 4-fluorophenyl boric acid
InChI Key LBUNNMJLXWQQBY-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(F)C=C1
IUPAC Name (4-fluorophenyl)boronic acid
Molecular Weight (g/mol) 139.92
PubChem CID 285645
ChEBI CHEBI:48661
Formula Weight 139.92
Physical Form Crystalline Powder
Show More Show Less