Learn More
4-Fluorophenylacetylene, 99%
CAS: 766-98-3 | C8H5F | 120.126 g/mol
Supplier: Thermo Scientific Chemicals H2626906
Description
Intermediates of liquid Crystals. As pharmaceutical intermediates. As organic synthesis intermediates. For vacuum deposition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIntermediates of liquid Crystals. As pharmaceutical intermediates. As organic synthesis intermediates. For vacuum deposition.
Solubility
Soluble in chloroform, diethyl ether, acetone, dichloromethane, hexane and ethanol. Highly soluble in toluene. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heavy metal powders.
Specifications
| 4-Fluorophenylacetylene | |
| 27°C to 28°C | |
| 55°C to 56°C (40 mmHg) | |
| Characteristic | |
| 1.517 | |
| 5 g | |
| 1099285 | |
| Soluble in chloroform,diethyl ether,acetone,dichloromethane,hexane and ethanol. Highly soluble in toluene. Insoluble in water. | |
| C#CC1=CC=C(C=C1)F | |
| 120.126 | |
| 120.12 |
| 766-98-3 | |
| 1.048 | |
| 29°C (85°F) | |
| C8H5F | |
| MFCD00168823 | |
| UN1325 | |
| 4-fluorophenylacetylene, benzene, 1-ethynyl-4-fluoro, 1-ethynyl-4-fluoro-benzene, 4'-fluorophenyl acetylene, p-fluorophenyl acetylene, 4'-fluorophenylacetylene, pubchem16719, 4-fluorophenyl acetylene, 4-fluoro phenylacetylene | |
| QXSWHQGIEKUBAS-UHFFFAOYSA-N | |
| 1-ethynyl-4-fluorobenzene | |
| 522627 | |
| 99% |
Chemical Identifiers
| 766-98-3 | |
| 120.126 | |
| QXSWHQGIEKUBAS-UHFFFAOYSA-N | |
| 522627 | |
| C#CC1=CC=C(C=C1)F |
| C8H5F | |
| MFCD00168823 | |
| 4-fluorophenylacetylene, benzene, 1-ethynyl-4-fluoro, 1-ethynyl-4-fluoro-benzene, 4'-fluorophenyl acetylene, p-fluorophenyl acetylene, 4'-fluorophenylacetylene, pubchem16719, 4-fluorophenyl acetylene, 4-fluoro phenylacetylene | |
| 1-ethynyl-4-fluorobenzene |
Safety and Handling
GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Hazard Class: 4.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only