missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetonitrile, 98+%

CAS: 459-22-3 | C8H6FN | 135.141 g/mol

Supplier:  Thermo Scientific Chemicals A1429022

 View more versions of this product

Catalog No. AAA1429022


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Fluorophenylacetonitrile
1.13
108°C (226°F)
1.5002
100 g
1907764
JHQBLYITVCBGTO-UHFFFAOYSA-N
2-(4-fluorophenyl)acetonitrile
68016
≥98%
459-22-3
238°C
C8H6FN
MFCD00001917
UN3276
4-fluorophenylacetonitrile, 4-fluorobenzyl cyanide, p-fluorobenzyl cyanide, benzeneacetonitrile, 4-fluoro, 4-fluorobenzylcyanide, 4-fluorobenzeneacetonitrile, p-fluorophenylacetonitrile, 2-4-fluorophenyl acetonitrile, 4-fluorobenzylnitrile, acetonitrile, p-fluorophenyl
C1=CC(=CC=C1CC#N)F
135.141
135.14

Chemical Identifiers

459-22-3
135.141
JHQBLYITVCBGTO-UHFFFAOYSA-N
68016
C1=CC(=CC=C1CC#N)F
C8H6FN
MFCD00001917
4-fluorophenylacetonitrile, 4-fluorobenzyl cyanide, p-fluorobenzyl cyanide, benzeneacetonitrile, 4-fluoro, 4-fluorobenzylcyanide, 4-fluorobenzeneacetonitrile, p-fluorophenylacetonitrile, 2-4-fluorophenyl acetonitrile, 4-fluorobenzylnitrile, acetonitrile, p-fluorophenyl
2-(4-fluorophenyl)acetonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 207-286-8

RTECSNumber : AM0210000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only