missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetone 98.0+%, TCI America™

Catalog No. p-7129738
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05355G 5 g
F053525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05355G Supplier TCI America Supplier No. F05355G
Only null left

Chemical Identifiers

CAS 459-03-0
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD00000362
InChI Key ZUEKIIWSVFBTCM-UHFFFAOYSA-N
Synonym 4-fluorophenylacetone, 4-fluorophenyl acetone, 1-4-fluorophenyl propan-2-one, 1-4-fluorophenyl acetone, 1-4-fluorophenyl-2-propanone, 2-propanone, 1-4-fluorophenyl, p-fluorophenyl acetone, 1-acetonyl-4-fluorobenzene, 1-4-fluoro-phenyl-propan-2-one
PubChem CID 521187
IUPAC Name 1-(4-fluorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(F)C=C1

Specifications

Boiling Point 130°C
Quantity 5 g
Formula Weight 152.17
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Fluorophenylacetone