missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetone, 99%

Catalog No. AAL1172006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1172006 5 g
AAL1172003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1172006 Supplier Thermo Scientific Chemicals Supplier No. L1172006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 459-03-0 | C9H9FO | 152.17 g/mol

4-Fluorophenylacetone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylacetone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 459-03-0
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD00000362
InChI Key ZUEKIIWSVFBTCM-UHFFFAOYSA-N
Synonym 4-fluorophenylacetone, 4-fluorophenyl acetone, 1-4-fluorophenyl propan-2-one, 1-4-fluorophenyl acetone, 1-4-fluorophenyl-2-propanone, 2-propanone, 1-4-fluorophenyl, p-fluorophenyl acetone, 1-acetonyl-4-fluorobenzene, 1-4-fluoro-phenyl-propan-2-one
PubChem CID 521187
IUPAC Name 1-(4-fluorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(F)C=C1

Specifications

Density 1.11
Boiling Point 106°C to 107°C (18 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.4958
Quantity 5 g
Beilstein 1936697
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 152.17
Percent Purity 99%
Chemical Name or Material 4-Fluorophenylacetone
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 207-284-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only