missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Fluorophenyl)urea 97.0+%, TCI America™
Supplier: TCI America F09505G
Specifications
| (4-Fluorophenyl)urea | |
| 180°C | |
| C7H7FN2O | |
| 5 g | |
| IQZBVVPYTDHTIP-UHFFFAOYSA-N | |
| (4-fluorophenyl)urea | |
| 12612 | |
| ≥97.0% (HPLC,N) |
| 659-30-3 | |
| White-Yellow | |
| MFCD00014787 | |
| 4-fluorophenyl urea, 1-4-fluorophenyl urea, urea, 1-p-fluorophenyl, 1-p-fluorophenyl urea, n-4-fluorophenyl urea, p-fluorophenylurea, amino-n-4-fluorophenyl amide, n'-4-fluorophenyl urea | |
| NC(=O)NC1=CC=C(F)C=C1 | |
| 154.14 | |
| 154.14 | |
| Crystalline Powder |
Chemical Identifiers
| 659-30-3 | |
| 154.14 | |
| IQZBVVPYTDHTIP-UHFFFAOYSA-N | |
| 12612 | |
| NC(=O)NC1=CC=C(F)C=C1 |
| C7H7FN2O | |
| MFCD00014787 | |
| 4-fluorophenyl urea, 1-4-fluorophenyl urea, urea, 1-p-fluorophenyl, 1-p-fluorophenyl urea, n-4-fluorophenyl urea, p-fluorophenylurea, amino-n-4-fluorophenyl amide, n'-4-fluorophenyl urea | |
| (4-fluorophenyl)urea |
Safety and Handling
RTECSNumber : YT3980000
TSCA : No