missing translation for 'onlineSavingsMsg'
Learn More

(4-Fluorophenyl)urea 97.0+%, TCI America™

Catalog No. F09501G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F09501G Supplier TCI America Supplier No. F09501G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 659-30-3
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.14
MDL Number MFCD00014787
InChI Key IQZBVVPYTDHTIP-UHFFFAOYSA-N
Synonym 4-fluorophenyl urea, 1-4-fluorophenyl urea, urea, 1-p-fluorophenyl, 1-p-fluorophenyl urea, n-4-fluorophenyl urea, p-fluorophenylurea, amino-n-4-fluorophenyl amide, n'-4-fluorophenyl urea
PubChem CID 12612
IUPAC Name (4-fluorophenyl)urea
SMILES NC(=O)NC1=CC=C(F)C=C1

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 1 g
Formula Weight 154.14
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4-Fluorophenyl)urea
RTECSNumber YT3980000
TSCA No