missing translation for 'onlineSavingsMsg'
Learn More

(4-Fluorophenyl)thiourea 97.0+%, TCI America™

Catalog No. p-7129929
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08365G 5 g
F083625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08365G Supplier TCI America Supplier No. F08365G
Only null left

Chemical Identifiers

CAS 459-05-2
Molecular Formula C7H7FN2S
Molecular Weight (g/mol) 170.21
MDL Number MFCD00041180
InChI Key BRWKXKNZRVALNZ-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl thiourea, 1-4-fluorophenyl-2-thiourea, 4-fluorophenyl thiourea, n-4-fluorophenyl thiourea, p-fluorophenylthiourea, 4-fluoro-phenyl-thiourea, thiourea, 4-fluorophenyl, amino 4-fluorophenyl amino methane-1-thione, pubchem10059, acmc-1ajjb
PubChem CID 693061
IUPAC Name (4-fluorophenyl)thiourea
SMILES NC(=S)NC1=CC=C(F)C=C1

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 170.21
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4-Fluorophenyl)thiourea