missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. F06445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06445G 5 g
F064425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06445G Supplier TCI America Supplier No. F06445G
Only null left

Chemical Identifiers

CAS 1544-68-9
Molecular Formula C7H4FNS
Molecular Weight (g/mol) 153.174
MDL Number MFCD00004809
InChI Key NFIUJHJMCQQYDL-UHFFFAOYSA-N
Synonym 4-fluorophenyl isothiocyanate, p-fluorophenyl isothiocyanate, benzene, 1-fluoro-4-isothiocyanato, 4-fluorophenylisothiocyanate, 1-fluoro-4-isothiocyanato-benzene, isothiocyanic acid, p-fluorophenyl ester, isothiocyanic acid 4-fluorophenyl ester, 4-fluorobenzenisothiocyanate, 4-fluoroisothiocyanatobenzene, pubchem5008
PubChem CID 15241
IUPAC Name 1-fluoro-4-isothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)F

Specifications

Color Yellow
Boiling Point 228°C
Quantity 5 g
UN Number 2922
Formula Weight 153.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorophenyl Isothiocyanate
RTECSNumber NX8760000
TSCA Yes
Recommended Storage Refrigerator