missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl Isocyanate 98.0+%, TCI America™

Catalog No. p-7131346
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
I03395G 5 g
I033925G 25 g
2 options

Catalog No. I03395G

Supplier: TCI America I03395G

Only null left

Chemical Identifiers

CAS 1195-45-5
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.113
MDL Number MFCD00002023
InChI Key DSVGFKBFFICWLZ-UHFFFAOYSA-N
PubChem CID 70955
IUPAC Name 1-fluoro-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)F

Specifications

Color Yellow
Boiling Point 68°C
Quantity 5 g
UN Number 2478
Synonym 4-fluorophenyl isocyanate, 4-fluorophenylisocyanate, p-fluorophenyl isocyanate, benzene, 1-fluoro-4-isocyanato, isocyanic acid 4-fluorophenyl ester, 1-fluoro-4-isocyanato-benzene, benzene,1-fluoro-4-isocyanato, 4-fluorobenzenisocyanate, paragos 331046, timtec-bb sbb032971
Formula Weight 137.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorophenyl Isocyanate
TSCA No
Recommended Storage Refrigerator