missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl isocyanate, 98+%

CAS: 1195-45-5 | C7H4FNO | 137.113 g/mol

$75.38 - $315.99

Chemical Identifiers

CAS 1195-45-5
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.113
MDL Number MFCD00002023
InChI Key DSVGFKBFFICWLZ-UHFFFAOYSA-N
PubChem CID 70955
IUPAC Name 1-fluoro-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1257606
View Documents
Thermo Scientific Chemicals
A1257606
5 g
Each for $75.38
Only null left
 
AAA1257614
View Documents
Thermo Scientific Chemicals
A1257614
25 g
Each for $315.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1195-45-5
137.113
DSVGFKBFFICWLZ-UHFFFAOYSA-N
1-fluoro-4-isocyanatobenzene
C7H4FNO
MFCD00002023
70955
C1=CC(=CC=C1N=C=O)F

Specifications

1195-45-5
55°C to 56°C (8 mmHg)
Pungent, Makes Eyes Water
1.514
5 g
471609
4-fluorophenyl isocyanate, 4-fluorophenylisocyanate, p-fluorophenyl isocyanate, benzene, 1-fluoro-4-isocyanato, isocyanic acid 4-fluorophenyl ester, 1-fluoro-4-isocyanato-benzene, benzene,1-fluoro-4-isocyanato, 4-fluorobenzenisocyanate, paragos 331046, timtec-bb sbb032971
C1=CC(=CC=C1N=C=O)F
137.113
137.11
4-Fluorophenyl isocyanate
1.21
52°C (125°F)
C7H4FNO
MFCD00002023
UN3080
Moisture sensitive; May form a precipitate
DSVGFKBFFICWLZ-UHFFFAOYSA-N
1-fluoro-4-isocyanatobenzene
70955
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H334-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P284-P285-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P363-P370+P378q-P501c

H226-H302-H312-H315-H317-H319-H330-H334-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.

EINECSNumber : 214-799-0

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only