missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl isocyanate, 98+%

Catalog No. AAA1257614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1257614 25 g
AAA1257606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1257614 Supplier Thermo Scientific Chemicals Supplier No. A1257614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1195-45-5 | C7H4FNO | 137.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1195-45-5
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.113
MDL Number MFCD00002023
InChI Key DSVGFKBFFICWLZ-UHFFFAOYSA-N
PubChem CID 70955
IUPAC Name 1-fluoro-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)F

Specifications

Density 1.21
Boiling Point 55°C to 56°C (8 mmHg)
Flash Point 52°C (125°F)
Odor Pungent, Makes Eyes Water
Refractive Index 1.514
Quantity 25 g
UN Number UN3080
Beilstein 471609
Sensitivity Moisture sensitive; May form a precipitate
Synonym 4-fluorophenyl isocyanate, 4-fluorophenylisocyanate, p-fluorophenyl isocyanate, benzene, 1-fluoro-4-isocyanato, isocyanic acid 4-fluorophenyl ester, 1-fluoro-4-isocyanato-benzene, benzene,1-fluoro-4-isocyanato, 4-fluorobenzenisocyanate, paragos 331046, timtec-bb sbb032971
Formula Weight 137.11
Percent Purity ≥98%
Chemical Name or Material 4-Fluorophenyl isocyanate
Show More Show Less
Hazard Category H226-H302-H312-H315-H317-H319-H330-H334-H335
Hazard Statement GHS H Statement
H300-H330-H334-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P284-P285-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
EINECSNumber 214-799-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only