missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenol 99.0+%, TCI America™

Catalog No. p-7129514
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
F016025G 25 g
F0160500G 500 g
2 options

Catalog No. F016025G

Supplier: TCI America F016025G

Only null left

Chemical Identifiers

CAS 371-41-5
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.10
MDL Number MFCD00002316
InChI Key RHMPLDJJXGPMEX-UHFFFAOYSA-N
Synonym p-fluorophenol, phenol, 4-fluoro, phenol, p-fluoro, para-fluorophenol, 4-fluoro phenol, 4-fluoro-phenol, ccris 665, 4-fluoranylphenol, fpn
PubChem CID 9732
IUPAC Name 4-fluorophenol
SMILES OC1=CC=C(F)C=C1

Specifications

Melting Point 47°C
Boiling Point 185°C
Quantity 25 g
Formula Weight 112.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorophenol
EINECSNumber (3)-4229
RTECSNumber SL4550000
TSCA Yes
Recommended Storage Refrigerator