missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenol, 99%

Catalog No. p-7081799
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1194514 25 g
AAA1194522 100 g
AAA1194536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1194514 Supplier Thermo Scientific Chemicals Supplier No. A1194514
Only null left

4-Fluorophenol, 99%, C6H5FO, CAS Number-371-41-5, ccris 665, fpn, 4-fluoro-phenol, p-fluorophenol, phenol, p-fluoro, para-fluorophenol, phenol, 4-fluoro, 4-fluoro phenol, 4-fluoranylphenol, 500g, 1362752, 185 deg.C to 188 deg.C, 1.22, Hazard Class: 6.1; Packaging Group: III, 206-736-0 | CAS: 371-41-5 | C6H5FO | 112.10 g/mol

4-Fluorophenol is a fluorinated phenolic compound with various applications as an starting reagent for the synthesis of pharmaceutical goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenol is a fluorinated phenolic compound with various applications as an starting reagent for the synthesis of pharmaceutical goods.

Solubility
Soluble in water (80 mg/ml at 20°C), methylene dichloride, chloroform and methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store away from strong bases and oxidizing agent.

Chemical Identifiers

CAS 371-41-5
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.10
MDL Number MFCD00002316
InChI Key RHMPLDJJXGPMEX-UHFFFAOYSA-N
Synonym p-fluorophenol, phenol, 4-fluoro, phenol, p-fluoro, para-fluorophenol, 4-fluoro phenol, 4-fluoro-phenol, ccris 665, 4-fluoranylphenol, fpn
PubChem CID 9732
IUPAC Name 4-fluorophenol
SMILES OC1=CC=C(F)C=C1

Specifications

Melting Point 44°C to 50°C
Density 1.22
Boiling Point 185°C to 188°C
Flash Point 68°C (154°F)
Odor Phenol-like
Quantity 25 g
UN Number UN2923
Beilstein 1362752
Solubility Information Soluble in water (80mg/ml at 20°C),methylene dichloride,chloroform and methanol.
Formula Weight 112.1
Percent Purity 99%
Chemical Name or Material 4-Fluorophenol
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 206-736-0
RTECSNumber SL4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only