missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoromandelic acid, 98%

Catalog No. p-7021164
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1339103 1 g
AAA1339106 5 g
AAA1339114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1339103 Supplier Thermo Scientific Chemicals Supplier No. A1339103
Only null left

CAS: 395-33-5 | C8H7FO3 | 170.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 395-33-5
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.14
MDL Number MFCD00044402,MFCD20638902
InChI Key RWCMOQXHIDWDDJ-UHFFFAOYNA-N
Synonym 4-fluoromandelic acid, 2-4-fluorophenyl-2-hydroxyacetic acid, p-fluoro mandelic acid, 4-fluorophenyl hydroxy acetic acid, p-fluoromandelic acid, 4-fluoro mandelic acid, 4-fluoromandelicacid, 4-fluoro-alpha-hydroxybenzeneacetic acid, 4-fluoro-dl-mandelic acid, 4-fluoro hydroxy acetic acid
PubChem CID 98068
IUPAC Name 2-(4-fluorophenyl)-2-hydroxyacetic acid
SMILES OC(C(O)=O)C1=CC=C(F)C=C1

Specifications

Melting Point 133°C to 138°C
Quantity 1 g
Beilstein 2211001
Formula Weight 170.14
Percent Purity 98%
Chemical Name or Material 4-Fluoromandelic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-899-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only