missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorodiphenyl ether, 99%

Catalog No. AAB2138022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2138022 100 g
AAB2138006 5 g
AAB2138014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2138022 Supplier Thermo Scientific Chemicals Supplier No. B2138022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 330-84-7 | C12H9FO | 188.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 330-84-7
Molecular Formula C12H9FO
Molecular Weight (g/mol) 188.20
MDL Number MFCD00055239
InChI Key AODSTUBSNYVSSL-UHFFFAOYSA-N
Synonym 4-fluorodiphenyl ether, 4-fluorophenyl phenyl ether, benzene, 1-fluoro-4-phenoxy, 4-fluoro-1-phenoxybenzene, 4-fluorodiphenylether, 4-fluorophenoxybenzene, pubchem1899, 4-fluoroduphenyl ether, 4-fluorophenylphenylether, #
PubChem CID 67614
IUPAC Name 1-fluoro-4-phenoxybenzene
SMILES FC1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Refractive Index 1.557
Quantity 100 g
Formula Weight 188.2
Percent Purity 99%
Chemical Name or Material 4-Fluorodiphenyl ether
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-357-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only