missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorocinnamic Acid 98.0+%, TCI America™
$59.69 - $173.26
Chemical Identifiers
| CAS | 459-32-5 |
|---|---|
| Molecular Formula | C9H7FO2 |
| Molecular Weight (g/mol) | 166.151 |
| MDL Number | MFCD00004395 |
| InChI Key | ISMMYAZSUSYVQG-ZZXKWVIFSA-N |
| Synonym | 4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e |
| PubChem CID | 1530234 |
| IUPAC Name | (E)-3-(4-fluorophenyl)prop-2-enoic acid |
| SMILES | C1=CC(=CC=C1C=CC(=O)O)F |
Chemical Identifiers
| 459-32-5 | |
| 166.151 | |
| ISMMYAZSUSYVQG-ZZXKWVIFSA-N | |
| 1530234 | |
| C1=CC(=CC=C1C=CC(=O)O)F |
| C9H7FO2 | |
| MFCD00004395 | |
| 4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e | |
| (E)-3-(4-fluorophenyl)prop-2-enoic acid |
Specifications
| 459-32-5 | |
| White-Yellow | |
| MFCD00004395 | |
| 4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e | |
| C1=CC(=CC=C1C=CC(=O)O)F | |
| 166.151 | |
| 166.15 | |
| Crystalline Powder |
| 207°C | |
| C9H7FO2 | |
| 5 g | |
| ISMMYAZSUSYVQG-ZZXKWVIFSA-N | |
| (E)-3-(4-fluorophenyl)prop-2-enoic acid | |
| 1530234 | |
| ≥98.0% (GC,T) | |
| 4-Fluorocinnamic Acid |
Safety and Handling
TSCA : No