missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorocinnamic Acid 98.0+%, TCI America™

Catalog No. f024425g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
F024425G 25 g
F02445G 5 g
2 options

Catalog No. F024425G

Supplier: TCI America F024425G

Only null left

Chemical Identifiers

CAS 459-32-5
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.151
MDL Number MFCD00004395
InChI Key ISMMYAZSUSYVQG-ZZXKWVIFSA-N
Synonym 4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e
PubChem CID 1530234
IUPAC Name (E)-3-(4-fluorophenyl)prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)F

Specifications

Melting Point 207°C
Color White-Yellow
Quantity 25 g
Formula Weight 166.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorocinnamic Acid