missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorocinnamaldehyde 95.0+%, TCI America™
Supplier: TCI America F072225G
Specifications
| 4-Fluorocinnamaldehyde | |
| Yellow | |
| C9H7FO | |
| 25 g | |
| YSIYEWBILJZDQH-OWOJBTEDSA-N | |
| (E)-3-(4-fluorophenyl)prop-2-enal | |
| 11217342 | |
| ≥95.0% (GC) |
| 24654-55-5 | |
| 95°C | |
| MFCD03425895 | |
| 3-(4-Fluorophenyl)acrylaldehyde, 3-(4-Fluorophenyl)-2-propenal | |
| C1=CC(=CC=C1C=CC=O)F | |
| 150.152 | |
| 150.15 | |
| Liquid |
Chemical Identifiers
| 24654-55-5 | |
| 150.152 | |
| YSIYEWBILJZDQH-OWOJBTEDSA-N | |
| 11217342 | |
| C1=CC(=CC=C1C=CC=O)F |
| C9H7FO | |
| MFCD03425895 | |
| 3-(4-Fluorophenyl)acrylaldehyde, 3-(4-Fluorophenyl)-2-propenal | |
| (E)-3-(4-fluorophenyl)prop-2-enal |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator