accessibility menu, dialog, popup

UserName

OF(E/Z)

CAS RN 24654-55-5

IUPAC Name: 3-(4-fluorophenyl)prop-2-enal
Synonyms: 3-(4-fluorophenyl)prop-2-enal
Molecular Weight (g/mol): 150.15
Molecular Formula: C9H7FO
InChi Key: YSIYEWBILJZDQH-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=CC=O)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results