missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorochalcone 98.0+%, TCI America™

Catalog No. F04595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04595G 5 g
F045925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04595G Supplier TCI America Supplier No. F04595G

Chemical Identifiers

CAS 1608-51-1
Molecular Formula C15H11FO
Molecular Weight (g/mol) 226.25
MDL Number MFCD00443467
InChI Key NYSCQZARWVHQBE-DHZHZOJOSA-N
Synonym 4-fluorochalcone, ccris 2224, 2e-3-4-fluorophenyl-1-phenylprop-2-en-1-one, 2-propen-1-one, 3-4-fluorophenyl-1-phenyl, e-3-4-fluorophenyl-1-phenyl-2-propen-1-one, e-3-4-fluorophenyl-1-phenyl-prop-2-en-1-one, 2-4-fluorobenzal acetophenone, 4-fluoro-chalcone, chalcone, 4-fluoro, 4-fluoro-trans-chalcone
PubChem CID 5366988
IUPAC Name (E)-3-(4-fluorophenyl)-1-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)F

Specifications

Melting Point 87°C
Color Yellow
Quantity 5 g
Formula Weight 226.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorochalcone