CAS RN 1608-51-1
IUPAC Name:
(2E)-3-(4-fluorophenyl)-1-phenylprop-2-en-1-one
Synonyms:
(2E)-3-(4-fluorophenyl)-1-phenylprop-2-en-1-one
Molecular Weight (g/mol):
226.25
Molecular Formula:
C15H11FO
InChi Key:
NYSCQZARWVHQBE-DHZHZOJOSA-N
SMILES:
FC1=CC=C(\C=C\C(=O)C2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
4-Fluorochalcone 98.0+%, TCI America™
CAS: 1608-51-1 Molecular Formula: C15H11FO Molecular Weight (g/mol): 226.25 MDL Number: MFCD00443467 InChI Key: NYSCQZARWVHQBE-DHZHZOJOSA-N Synonym: 4-fluorochalcone,ccris 2224,2e-3-4-fluorophenyl-1-phenylprop-2-en-1-one,2-propen-1-one, 3-4-fluorophenyl-1-phenyl,e-3-4-fluorophenyl-1-phenyl-2-propen-1-one,e-3-4-fluorophenyl-1-phenyl-prop-2-en-1-one,2-4-fluorobenzal acetophenone,4-fluoro-chalcone,chalcone, 4-fluoro,4-fluoro-trans-chalcone PubChem CID: 5366988 IUPAC Name: (E)-3-(4-fluorophenyl)-1-phenylprop-2-en-1-one SMILES: C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)F