missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluorobiphenyl-4-carboxylic acid, 96%

CAS: 5731-10-2 | C13H8FO2 | 215.20 g/mol

$563.14 - $563.14

Chemical Identifiers

CAS 5731-10-2
Molecular Formula C13H8FO2
Molecular Weight (g/mol) 215.20
MDL Number MFCD01631909
InChI Key LXWNTLBMNCXRQN-UHFFFAOYSA-M
Synonym 4-4-fluorophenyl benzoic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4'-fluorobiphenyl-4-carboxylic acid, 4'-fluoro-1,1'-biphenyl-4-carboxylic acid, 4-biphenyl-4'-fluoro-carboxylic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4-fluoro-biphenyl-4-carboxylic acid, 4'-fluoro 1,1'-biphenyl-4-carboxylic acid, 4fc
PubChem CID 2782689
IUPAC Name 4-(4-fluorophenyl)benzoic acid
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)F)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3320806
View Documents
Thermo Scientific Chemicals
H3320806
5 g
Each for $563.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5731-10-2
215.20
LXWNTLBMNCXRQN-UHFFFAOYSA-M
2782689
C1=CC(=CC=C1C2=CC=C(C=C2)F)C(=O)O
C13H8FO2
MFCD01631909
4-4-fluorophenyl benzoic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4'-fluorobiphenyl-4-carboxylic acid, 4'-fluoro-1,1'-biphenyl-4-carboxylic acid, 4-biphenyl-4'-fluoro-carboxylic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4-fluoro-biphenyl-4-carboxylic acid, 4'-fluoro 1,1'-biphenyl-4-carboxylic acid, 4fc
4-(4-fluorophenyl)benzoic acid

Specifications

5731-10-2
C13H8FO2
5 g
LXWNTLBMNCXRQN-UHFFFAOYSA-M
4-(4-fluorophenyl)benzoic acid
2782689
96%
243°C to 244°C
MFCD01631909
4-4-fluorophenyl benzoic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4'-fluorobiphenyl-4-carboxylic acid, 4'-fluoro-1,1'-biphenyl-4-carboxylic acid, 4-biphenyl-4'-fluoro-carboxylic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4-fluoro-biphenyl-4-carboxylic acid, 4'-fluoro 1,1'-biphenyl-4-carboxylic acid, 4fc
C1=CC(=CC=C1C2=CC=C(C=C2)F)C(=O)O
215.20
216.21
4'-Fluorobiphenyl-4-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only