missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzylamine 98.0+%, TCI America™
Supplier: TCI America F025225G
Specifications
| 4-Fluorobenzylamine | |
| Yellow | |
| C7H8FN | |
| 25 g | |
| 4-fluorobenzylamine, 4-fluorophenyl methanamine, benzenemethanamine, 4-fluoro, p-fluorobenzylamine, benzylamine, p-fluoro, p-fluorobenzyl amine, 4-fluoro-benzylamine, 1-4-fluorophenyl methanamine, 4-fluorobenzyl amine, chembl12392 | |
| C1=CC(=CC=C1CN)F | |
| 125.146 | |
| 125.15 | |
| Liquid |
| 140-75-0 | |
| 183°C | |
| MFCD00008120 | |
| 2735 | |
| IIFVWLUQBAIPMJ-UHFFFAOYSA-N | |
| (4-fluorophenyl)methanamine | |
| 67326 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 140-75-0 | |
| 125.146 | |
| IIFVWLUQBAIPMJ-UHFFFAOYSA-N | |
| 67326 | |
| C1=CC(=CC=C1CN)F |
| C7H8FN | |
| MFCD00008120 | |
| 4-fluorobenzylamine, 4-fluorophenyl methanamine, benzenemethanamine, 4-fluoro, p-fluorobenzylamine, benzylamine, p-fluoro, p-fluorobenzyl amine, 4-fluoro-benzylamine, 1-4-fluorophenyl methanamine, 4-fluorobenzyl amine, chembl12392 | |
| (4-fluorophenyl)methanamine |
Safety and Handling
TSCA : Yes