Learn More
4-Fluorobenzylamine, 98+%
CAS: 140-75-0 | C7H8FN | 125.146 g/mol
Supplier: Thermo Scientific Chemicals A1548618
Description
4-Fluorobenzylamine is used as a potassium channel blocking agent. It is used in the synthesis of new tris-iron(III) chelates of 3-hydroxy-4-pyridinone ligands. It reacts with the α- and γ-carboxyl groups of folic acid to yield 18F-labeled folate2. It is an important building block for the synthesis of 18F-labeled compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluorobenzylamine is used as a potassium channel blocking agent. It is used in the synthesis of new tris-iron(III) chelates of 3-hydroxy-4-pyridinone ligands. It reacts with the α- and γ-carboxyl groups of folic acid to yield 18F-labeled folate2. It is an important building block for the synthesis of 18F-labeled compounds.
Solubility
Soluble in water.
Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agent, air, carbon dioxide.
Specifications
| 4-Fluorobenzylamine | |
| 1.095 | |
| 66°C (150°F) | |
| C7H8FN | |
| MFCD00008120 | |
| UN2735 | |
| Air Sensitive | |
| Soluble in water. | |
| C1=CC(=CC=C1CN)F | |
| 125.146 | |
| 125.15 |
| 140-75-0 | |
| 182°C to 183°C | |
| Amine-like | |
| 1.513 | |
| 50 g | |
| 636501 | |
| 4-fluorobenzylamine, 4-fluorophenyl methanamine, benzenemethanamine, 4-fluoro, p-fluorobenzylamine, benzylamine, p-fluoro, p-fluorobenzyl amine, 4-fluoro-benzylamine, 1-4-fluorophenyl methanamine, 4-fluorobenzyl amine, chembl12392 | |
| IIFVWLUQBAIPMJ-UHFFFAOYSA-N | |
| (4-fluorophenyl)methanamine | |
| 67326 | |
| ≥98% |
Chemical Identifiers
| 140-75-0 | |
| 125.146 | |
| IIFVWLUQBAIPMJ-UHFFFAOYSA-N | |
| 67326 | |
| C1=CC(=CC=C1CN)F |
| C7H8FN | |
| MFCD00008120 | |
| 4-fluorobenzylamine, 4-fluorophenyl methanamine, benzenemethanamine, 4-fluoro, p-fluorobenzylamine, benzylamine, p-fluoro, p-fluorobenzyl amine, 4-fluoro-benzylamine, 1-4-fluorophenyl methanamine, 4-fluorobenzyl amine, chembl12392 | |
| (4-fluorophenyl)methanamine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 205-430-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only