missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzylamine, 97%

Catalog No. p-4510562
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC171950250 25 g Glass bottle
AC171951000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC171950250 Supplier Thermo Scientific Chemicals Supplier No. 171950250

CAS: 140-75-0 | C7H8FN | 125.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140-75-0
InChI Key IIFVWLUQBAIPMJ-UHFFFAOYSA-N
Synonym 4-fluorobenzylamine, 4-fluorophenyl methanamine, benzenemethanamine, 4-fluoro, p-fluorobenzylamine, benzylamine, p-fluoro, p-fluorobenzyl amine, 4-fluoro-benzylamine, 1-4-fluorophenyl methanamine, 4-fluorobenzyl amine, chembl12392
PubChem CID 67326
IUPAC Name (4-fluorophenyl)methanamine
SMILES C1=CC(=CC=C1CN)F

Specifications

Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 183.0°C to 185.0°C
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H8FN
Refractive Index 1.5117
Linear Formula FC6H4CH2NH2
Quantity 25 g
Specific Gravity 1.09
Molecular Weight (g/mol) 125.15
Formula Weight 125.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 205-430-4

RUO – Research Use Only