missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl mercaptan, 96%

Catalog No. p-7080559
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1012603 1 g
AAA1012606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1012603 Supplier Thermo Scientific Chemicals Supplier No. A1012603

CAS: 15894-04-9 | C7H7FS | 142.191 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15894-04-9
Molecular Formula C7H7FS
Molecular Weight (g/mol) 142.191
MDL Number MFCD00040724
InChI Key RKTRHMNWVZRZJQ-UHFFFAOYSA-N
Synonym 4-fluorobenzyl mercaptan, 4-fluorophenyl methanethiol, 4-fluorobenzylmercaptan, 4-fluoro benzyl mercaptan, 4-fluorobenzenemethanethiol, p-fluorotoluene-alpha-thiol, benzenemethanethiol, 4-fluoro, 4-fluoro-phenyl-methanethiol, 4-fluorobenzylthiol, 4-fluorobenzyl thiol
PubChem CID 85171
IUPAC Name (4-fluorophenyl)methanethiol
SMILES C1=CC(=CC=C1CS)F

Specifications

Density 1.157
Boiling Point 72°C to 74°C (15 mmHg)
Flash Point 76°C (168°F)
Refractive Index 1.543
Quantity 1 g
UN Number UN2810
Beilstein 2242520
Sensitivity Air Sensitive
Formula Weight 142.2
Percent Purity 96%
Chemical Name or Material 4-Fluorobenzyl mercaptan
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 240-031-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only