missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl isocyanate, 98%

Catalog No. AC319300010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC319300010 1 g Glass bottle
AC319300050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC319300010 Supplier Thermo Scientific Chemicals Supplier No. 319300010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 132740-43-3 | C8H6FNO | 151.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132740-43-3
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD00673062
InChI Key HHSIWJYERNCLKQ-UHFFFAOYSA-N
Synonym 4-fluorobenzyl isocyanate, 1-fluoro-4-isocyanatomethyl benzene, 4-fluorobenzylisocyanate, 4-fluorophenyl methanisocyanate, 4-fluorobezylisocyanate, acmc-1c0to, intermediates-zcf02017, 4-fluoro benzyl isocyanate, 4-fluorophenyl methyl isocyanate
PubChem CID 2733376
IUPAC Name 1-fluoro-4-(isocyanatomethyl)benzene
SMILES FC1=CC=C(CN=C=O)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1860g/mL
Boiling Point 211.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5040 to 1.5060
Linear Formula FC6H4CH2NCO
Quantity 1 g
Specific Gravity 1.186
Formula Weight 151.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only