missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzyl Cyanide 98.0+%, TCI America™
$59.69 - $352.31
Chemical Identifiers
| CAS | 459-22-3 |
|---|---|
| Molecular Formula | C8H6FN |
| Molecular Weight (g/mol) | 135.141 |
| MDL Number | MFCD00001917 |
| InChI Key | JHQBLYITVCBGTO-UHFFFAOYSA-N |
| Synonym | 4-fluorophenylacetonitrile, 4-fluorobenzyl cyanide, p-fluorobenzyl cyanide, benzeneacetonitrile, 4-fluoro, 4-fluorobenzylcyanide, 4-fluorobenzeneacetonitrile, p-fluorophenylacetonitrile, 2-4-fluorophenyl acetonitrile, 4-fluorobenzylnitrile, acetonitrile, p-fluorophenyl |
| PubChem CID | 68016 |
| IUPAC Name | 2-(4-fluorophenyl)acetonitrile |
| SMILES | C1=CC(=CC=C1CC#N)F |
Chemical Identifiers
| 459-22-3 | |
| 135.141 | |
| JHQBLYITVCBGTO-UHFFFAOYSA-N | |
| 68016 | |
| C1=CC(=CC=C1CC#N)F |
| C8H6FN | |
| MFCD00001917 | |
| 4-fluorophenylacetonitrile, 4-fluorobenzyl cyanide, p-fluorobenzyl cyanide, benzeneacetonitrile, 4-fluoro, 4-fluorobenzylcyanide, 4-fluorobenzeneacetonitrile, p-fluorophenylacetonitrile, 2-4-fluorophenyl acetonitrile, 4-fluorobenzylnitrile, acetonitrile, p-fluorophenyl | |
| 2-(4-fluorophenyl)acetonitrile |
Specifications
| 459-22-3 | |
| 230°C | |
| MFCD00001917 | |
| 3276 | |
| JHQBLYITVCBGTO-UHFFFAOYSA-N | |
| 2-(4-fluorophenyl)acetonitrile | |
| 68016 | |
| ≥98.0% (GC) | |
| 4-Fluorobenzyl Cyanide |
| Yellow | |
| C8H6FN | |
| 25 g | |
| 4-fluorophenylacetonitrile, 4-fluorobenzyl cyanide, p-fluorobenzyl cyanide, benzeneacetonitrile, 4-fluoro, 4-fluorobenzylcyanide, 4-fluorobenzeneacetonitrile, p-fluorophenylacetonitrile, 2-4-fluorophenyl acetonitrile, 4-fluorobenzylnitrile, acetonitrile, p-fluorophenyl | |
| C1=CC(=CC=C1CC#N)F | |
| 135.141 | |
| 135.14 | |
| Liquid |
Safety and Handling
RTECSNumber : AM0210000
TSCA : Yes