missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzyl Chloride 98.0+%, TCI America™
$134.75 - $852.85
Chemical Identifiers
| CAS | 352-11-4 |
|---|---|
| Molecular Formula | C7H6ClF |
| Molecular Weight (g/mol) | 144.573 |
| MDL Number | MFCD00000913 |
| InChI Key | IZXWCDITFDNEBY-UHFFFAOYSA-N |
| Synonym | 4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene |
| PubChem CID | 9602 |
| IUPAC Name | 1-(chloromethyl)-4-fluorobenzene |
| SMILES | C1=CC(=CC=C1CCl)F |
Chemical Identifiers
| 352-11-4 | |
| 144.573 | |
| IZXWCDITFDNEBY-UHFFFAOYSA-N | |
| 9602 | |
| C1=CC(=CC=C1CCl)F |
| C7H6ClF | |
| MFCD00000913 | |
| 4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene | |
| 1-(chloromethyl)-4-fluorobenzene |
Specifications
| 352-11-4 | |
| 185°C | |
| MFCD00000913 | |
| 2920 | |
| IZXWCDITFDNEBY-UHFFFAOYSA-N | |
| 1-(chloromethyl)-4-fluorobenzene | |
| 9602 | |
| ≥98.0% (GC) | |
| 4-Fluorobenzyl Chloride |
| Colorless | |
| C7H6ClF | |
| 25 g | |
| 4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene | |
| C1=CC(=CC=C1CCl)F | |
| 144.573 | |
| 144.57 | |
| Liquid |
Safety and Handling
TSCA : Yes