missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl Chloride 98.0+%, TCI America™

Catalog No. F0204500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0204500G 500 g
F020425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0204500G Supplier TCI America Supplier No. F0204500G

Chemical Identifiers

CAS 352-11-4
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.573
MDL Number MFCD00000913
InChI Key IZXWCDITFDNEBY-UHFFFAOYSA-N
Synonym 4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene
PubChem CID 9602
IUPAC Name 1-(chloromethyl)-4-fluorobenzene
SMILES C1=CC(=CC=C1CCl)F

Specifications

Color Colorless
Boiling Point 185°C
Quantity 500 g
UN Number 2920
Formula Weight 144.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzyl Chloride