missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl chloride, 99%

CAS: 352-11-4 | C7H6ClF | 144.573 g/mol

Supplier:  Thermo Scientific Chemicals A1468622

 View more versions of this product

Catalog No. AAA1468622

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Fluorobenzyl chloride
-18°C
185°C
Pungent
1.513
100 g
742272
IZXWCDITFDNEBY-UHFFFAOYSA-N
1-(chloromethyl)-4-fluorobenzene
9602
99%
352-11-4
1.214
69°C (156°F)
C7H6ClF
MFCD00000913
UN3265
4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene
C1=CC(=CC=C1CCl)F
144.573
144.58

Chemical Identifiers

352-11-4
144.573
IZXWCDITFDNEBY-UHFFFAOYSA-N
9602
C1=CC(=CC=C1CCl)F
C7H6ClF
MFCD00000913
4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene
1-(chloromethyl)-4-fluorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 206-516-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only