missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl chloride, 99%

Catalog No. AAA1468636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1468636 500 g
AAA1468614 25 g
AAA1468622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1468636 Supplier Thermo Scientific Chemicals Supplier No. A1468636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 352-11-4 | C7H6ClF | 144.573 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 352-11-4
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.573
MDL Number MFCD00000913
InChI Key IZXWCDITFDNEBY-UHFFFAOYSA-N
Synonym 4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene
PubChem CID 9602
IUPAC Name 1-(chloromethyl)-4-fluorobenzene
SMILES C1=CC(=CC=C1CCl)F

Specifications

Melting Point -18°C
Density 1.214
Boiling Point 185°C
Flash Point 69°C (156°F)
Odor Pungent
Refractive Index 1.513
Quantity 500 g
UN Number UN3265
Beilstein 742272
Formula Weight 144.58
Percent Purity 99%
Chemical Name or Material 4-Fluorobenzyl chloride
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-516-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only