missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl bromide, 97%

CAS: 459-46-1 | C7H6BrF | 189.03 g/mol

$97.97 - $1615.26

Chemical Identifiers

CAS 459-46-1
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.03
MDL Number MFCD00000359
InChI Key NVNPLEPBDPJYRZ-UHFFFAOYSA-N
Synonym 4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide
PubChem CID 68021
IUPAC Name 1-(bromomethyl)-4-fluorobenzene
SMILES FC1=CC=C(CBr)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1020809
View Documents
Thermo Scientific Chemicals
A1020809
10 g
Each for $97.97
Only null left
 
AAA1020818
View Documents
Thermo Scientific Chemicals
A1020818
50 g
Each for $384.84
Only null left
 
AAA1020830
View Documents
Thermo Scientific Chemicals
A1020830
250 g
Each for $1,615.26
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

459-46-1
189.03
NVNPLEPBDPJYRZ-UHFFFAOYSA-N
68021
FC1=CC=C(CBr)C=C1
C7H6BrF
MFCD00000359
4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide
1-(bromomethyl)-4-fluorobenzene

Specifications

459-46-1
84°C to 85°C (15 mmHg)
Pungent
1.547
10 g
636507
NVNPLEPBDPJYRZ-UHFFFAOYSA-N
1-(bromomethyl)-4-fluorobenzene
68021
97%
1.53
66°C (150°F)
C7H6BrF
MFCD00000359
UN3265
4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide
FC1=CC=C(CBr)C=C1
189.03
189.03
4-Fluorobenzyl bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 207-291-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only