missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzoyl Chloride 97.0+%, TCI America™
$76.43 - $400.78
Chemical Identifiers
| CAS | 403-43-0 |
|---|---|
| Molecular Formula | C7H4ClFO |
| Molecular Weight (g/mol) | 158.556 |
| MDL Number | MFCD00000684 |
| InChI Key | CZKLEJHVLCMVQR-UHFFFAOYSA-N |
| Synonym | benzoyl chloride, 4-fluoro, p-fluorobenzoyl chloride, benzoyl chloride, p-fluoro, p-fluorobenzoic acid chloride, 4-fluorobenzoylchloride, fluorobenzoyl 4-chloride, 4-fluorobenzoic acid chloride, p-fluorobenzoylchloride, p-fluorobenzoyl choride, 4-flurobenzoyl chloride |
| PubChem CID | 67879 |
| IUPAC Name | 4-fluorobenzoyl chloride |
| SMILES | C1=CC(=CC=C1C(=O)Cl)F |
Chemical Identifiers
| 403-43-0 | |
| 158.556 | |
| CZKLEJHVLCMVQR-UHFFFAOYSA-N | |
| 67879 | |
| C1=CC(=CC=C1C(=O)Cl)F |
| C7H4ClFO | |
| MFCD00000684 | |
| benzoyl chloride, 4-fluoro, p-fluorobenzoyl chloride, benzoyl chloride, p-fluoro, p-fluorobenzoic acid chloride, 4-fluorobenzoylchloride, fluorobenzoyl 4-chloride, 4-fluorobenzoic acid chloride, p-fluorobenzoylchloride, p-fluorobenzoyl choride, 4-flurobenzoyl chloride | |
| 4-fluorobenzoyl chloride |
Specifications
| 403-43-0 | |
| 193°C | |
| MFCD00000684 | |
| 3265 | |
| CZKLEJHVLCMVQR-UHFFFAOYSA-N | |
| 4-fluorobenzoyl chloride | |
| 67879 | |
| ≥97.0% (T) | |
| 4-Fluorobenzoyl Chloride |
| Yellow | |
| C7H4ClFO | |
| 25 g | |
| benzoyl chloride, 4-fluoro, p-fluorobenzoyl chloride, benzoyl chloride, p-fluoro, p-fluorobenzoic acid chloride, 4-fluorobenzoylchloride, fluorobenzoyl 4-chloride, 4-fluorobenzoic acid chloride, p-fluorobenzoylchloride, p-fluorobenzoyl choride, 4-flurobenzoyl chloride | |
| C1=CC(=CC=C1C(=O)Cl)F | |
| 158.556 | |
| 158.56 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-4467
TSCA : Yes