missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoyl Chloride 97.0+%, TCI America™

Catalog No. F0213500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0213500G 500 g
F021325G 25 g
F0213100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F0213500G Supplier TCI America Supplier No. F0213500G

Chemical Identifiers

CAS 403-43-0
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00000684
InChI Key CZKLEJHVLCMVQR-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-fluoro, p-fluorobenzoyl chloride, benzoyl chloride, p-fluoro, p-fluorobenzoic acid chloride, 4-fluorobenzoylchloride, fluorobenzoyl 4-chloride, 4-fluorobenzoic acid chloride, p-fluorobenzoylchloride, p-fluorobenzoyl choride, 4-flurobenzoyl chloride
PubChem CID 67879
IUPAC Name 4-fluorobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)F

Specifications

Color Yellow
Boiling Point 193°C
Quantity 500 g
UN Number 3265
Formula Weight 158.56
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzoyl Chloride
EINECSNumber (3)-4467
TSCA Yes