missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoyl chloride, 98%

CAS: 403-43-0 | C7H4ClFO | 158.556 g/mol

$91.44 - $1226.89

Chemical Identifiers

CAS 403-43-0
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00000684
InChI Key CZKLEJHVLCMVQR-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-fluoro, p-fluorobenzoyl chloride, benzoyl chloride, p-fluoro, p-fluorobenzoic acid chloride, 4-fluorobenzoylchloride, fluorobenzoyl 4-chloride, 4-fluorobenzoic acid chloride, p-fluorobenzoylchloride, p-fluorobenzoyl choride, 4-flurobenzoyl chloride
PubChem CID 67879
IUPAC Name 4-fluorobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1294614
View Documents
Thermo Scientific Chemicals
A1294614
25 g
Each for $91.44
Only null left
 
AAA1294622
View Documents
Thermo Scientific Chemicals
A1294622
100 g
Each for $254.21
Only null left
 
AAA1294636
View Documents
Thermo Scientific Chemicals
A1294636
500 g
Each for $1,226.89
Only null left
 
Description

Description

It is used in the synthesis of polyphenylene ether and thioether ketones and also in the synthesis fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties. It is also used as a raw material in engineering of plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of polyphenylene ether and thioether ketones and also in the synthesis fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties. It is also used as a raw material in engineering of plastics.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under gas.
Specifications

Chemical Identifiers

403-43-0
158.556
CZKLEJHVLCMVQR-UHFFFAOYSA-N
67879
C1=CC(=CC=C1C(=O)Cl)F
C7H4ClFO
MFCD00000684
benzoyl chloride, 4-fluoro, p-fluorobenzoyl chloride, benzoyl chloride, p-fluoro, p-fluorobenzoic acid chloride, 4-fluorobenzoylchloride, fluorobenzoyl 4-chloride, 4-fluorobenzoic acid chloride, p-fluorobenzoylchloride, p-fluorobenzoyl choride, 4-flurobenzoyl chloride
4-fluorobenzoyl chloride

Specifications

403-43-0
1.326
82°C (179°F)
C7H4ClFO
MFCD00000684
UN3265
Moisture sensitive
Reacts with water.
C1=CC(=CC=C1C(=O)Cl)F
158.556
158.56
4-Fluorobenzoyl chloride
10°C to 12°C
193°C
Acidic
1.532
25 g
386215
benzoyl chloride, 4-fluoro, p-fluorobenzoyl chloride, benzoyl chloride, p-fluoro, p-fluorobenzoic acid chloride, 4-fluorobenzoylchloride, fluorobenzoyl 4-chloride, 4-fluorobenzoic acid chloride, p-fluorobenzoylchloride, p-fluorobenzoyl choride, 4-flurobenzoyl chloride
CZKLEJHVLCMVQR-UHFFFAOYSA-N
4-fluorobenzoyl chloride
67879
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 206-961-4

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only