missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoyl chloride, 98%

Catalog No. AAA1294636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1294636 500 g
AAA1294614 25 g
AAA1294622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1294636 Supplier Thermo Scientific Chemicals Supplier No. A1294636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 403-43-0 | C7H4ClFO | 158.556 g/mol

It is used in the synthesis of polyphenylene ether and thioether ketones and also in the synthesis fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties. It is also used as a raw material in engineering of plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of polyphenylene ether and thioether ketones and also in the synthesis fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties. It is also used as a raw material in engineering of plastics.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under gas.

Chemical Identifiers

CAS 403-43-0
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00000684
InChI Key CZKLEJHVLCMVQR-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-fluoro, p-fluorobenzoyl chloride, benzoyl chloride, p-fluoro, p-fluorobenzoic acid chloride, 4-fluorobenzoylchloride, fluorobenzoyl 4-chloride, 4-fluorobenzoic acid chloride, p-fluorobenzoylchloride, p-fluorobenzoyl choride, 4-flurobenzoyl chloride
PubChem CID 67879
IUPAC Name 4-fluorobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)F

Specifications

Melting Point 10°C to 12°C
Density 1.326
Boiling Point 193°C
Flash Point 82°C (179°F)
Odor Acidic
Refractive Index 1.532
Quantity 500 g
UN Number UN3265
Beilstein 386215
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 158.56
Percent Purity 98%
Chemical Name or Material 4-Fluorobenzoyl chloride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-961-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only