Learn More
4-Fluorobenzotrifluoride, 98+%
CAS: 402-44-8 | C7H4F4 | 164.103 g/mol
$88.99 - $284.22
Chemical Identifiers
| CAS | 402-44-8 |
|---|---|
| Molecular Formula | C7H4F4 |
| Molecular Weight (g/mol) | 164.103 |
| MDL Number | MFCD00000399 |
| InChI Key | UNNNAIWPDLRVRN-UHFFFAOYSA-N |
| Synonym | 4-fluorobenzotrifluoride, 1-fluoro-4-trifluoromethyl benzene, p-fluorobenzotrifluoride, benzene, 1-fluoro-4-trifluoromethyl, alpha,alpha,alpha,4-tetrafluorotoluene, p-fluorotrifluoromethylbenzene, p-trifluoromethyl fluorobenzene, 4-flourobenzotrifluoride, a,a,a,4-tetrafluorotoluene, 4-trifluoromethyl fluorobenzene |
| PubChem CID | 67873 |
| IUPAC Name | 1-fluoro-4-(trifluoromethyl)benzene |
| SMILES | C1=CC(=CC=C1C(F)(F)F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1337706
|
Thermo Scientific Chemicals
A1337706 |
5 g |
Each for $88.99
|
|
|||||
|
AAA1337714
|
Thermo Scientific Chemicals
A1337714 |
25 g |
Each for $284.22
|
|
|||||
Description
4-Fluorobenzotrifluoride was used in the synthesis of 2,2?-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2?-biphenol, (S)-fluoxetine, potent inhibitor of neuronal serotonin-uptake and 1-aryloxy-2-substituted aminomethyltetrahydronaphthalene derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluorobenzotrifluoride was used in the synthesis of 2,2′-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2′-biphenol, (S)-fluoxetine, potent inhibitor of neuronal serotonin-uptake and 1-aryloxy-2-substituted aminomethyltetrahydronaphthalene derivatives.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Chemical Identifiers
| 402-44-8 | |
| 164.103 | |
| UNNNAIWPDLRVRN-UHFFFAOYSA-N | |
| 67873 | |
| C1=CC(=CC=C1C(F)(F)F)F |
| C7H4F4 | |
| MFCD00000399 | |
| 4-fluorobenzotrifluoride, 1-fluoro-4-trifluoromethyl benzene, p-fluorobenzotrifluoride, benzene, 1-fluoro-4-trifluoromethyl, alpha,alpha,alpha,4-tetrafluorotoluene, p-fluorotrifluoromethylbenzene, p-trifluoromethyl fluorobenzene, 4-flourobenzotrifluoride, a,a,a,4-tetrafluorotoluene, 4-trifluoromethyl fluorobenzene | |
| 1-fluoro-4-(trifluoromethyl)benzene |
Specifications
| 402-44-8 | |
| 1.295 | |
| 10°C (50°F) | |
| 1.401 | |
| 5 g | |
| 1909390 | |
| Slightly soluble in water. | |
| C1=CC(=CC=C1C(F)(F)F)F | |
| 164.103 | |
| 164.1 | |
| 4-Fluorobenzotrifluoride |
| -42°C | |
| 103°C to 105°C | |
| C7H4F4 | |
| MFCD00000399 | |
| UN1993 | |
| 4-fluorobenzotrifluoride, 1-fluoro-4-trifluoromethyl benzene, p-fluorobenzotrifluoride, benzene, 1-fluoro-4-trifluoromethyl, alpha,alpha,alpha,4-tetrafluorotoluene, p-fluorotrifluoromethylbenzene, p-trifluoromethyl fluorobenzene, 4-flourobenzotrifluoride, a,a,a,4-tetrafluorotoluene, 4-trifluoromethyl fluorobenzene | |
| UNNNAIWPDLRVRN-UHFFFAOYSA-N | |
| 1-fluoro-4-(trifluoromethyl)benzene | |
| 67873 | |
| ≥98% |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 206-944-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only