missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzotrifluoride, 98+%

Catalog No. AAA1337706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1337706 5 g
AAA1337714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1337706 Supplier Thermo Scientific Chemicals Supplier No. A1337706

CAS: 402-44-8 | C7H4F4 | 164.103 g/mol

4-Fluorobenzotrifluoride was used in the synthesis of 2,2?-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2?-biphenol, (S)-fluoxetine, potent inhibitor of neuronal serotonin-uptake and 1-aryloxy-2-substituted aminomethyltetrahydronaphthalene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzotrifluoride was used in the synthesis of 2,2′-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2′-biphenol, (S)-fluoxetine, potent inhibitor of neuronal serotonin-uptake and 1-aryloxy-2-substituted aminomethyltetrahydronaphthalene derivatives.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 402-44-8
Molecular Formula C7H4F4
Molecular Weight (g/mol) 164.103
MDL Number MFCD00000399
InChI Key UNNNAIWPDLRVRN-UHFFFAOYSA-N
Synonym 4-fluorobenzotrifluoride, 1-fluoro-4-trifluoromethyl benzene, p-fluorobenzotrifluoride, benzene, 1-fluoro-4-trifluoromethyl, alpha,alpha,alpha,4-tetrafluorotoluene, p-fluorotrifluoromethylbenzene, p-trifluoromethyl fluorobenzene, 4-flourobenzotrifluoride, a,a,a,4-tetrafluorotoluene, 4-trifluoromethyl fluorobenzene
PubChem CID 67873
IUPAC Name 1-fluoro-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)F

Specifications

Melting Point -42°C
Density 1.295
Boiling Point 103°C to 105°C
Flash Point 10°C (50°F)
Refractive Index 1.401
Quantity 5 g
UN Number UN1993
Beilstein 1909390
Solubility Information Slightly soluble in water.
Formula Weight 164.1
Percent Purity ≥98%
Chemical Name or Material 4-Fluorobenzotrifluoride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-944-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only