missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzonitrile, 99%

Catalog No. AC160810050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160810050 5 g Glass bottle
AC160810250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC160810050 Supplier Thermo Scientific Chemicals Supplier No. 160810050

CAS: 1194-02-1 | C7H4FN | 121.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1194-02-1
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001812
InChI Key AEKVBBNGWBBYLL-UHFFFAOYSA-N
Synonym p-fluorobenzonitrile, benzonitrile, 4-fluoro, 4-cyanofluorobenzene, p-cyanofluorobenzene, benzonitrile, p-fluoro, 4-fluoro-benzonitrile, 4-fluorobenzenecarbonitrile, 4-fluorobenzonitril, pfbn, 1-cyano-4-fluorobenzene
PubChem CID 14517
IUPAC Name 4-fluorobenzonitrile
SMILES C1=CC(=CC=C1C#N)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 33°C to 36°C
Color White
Boiling Point 188°C (750.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CN
Quantity 5 g
Beilstein 09, I, 138
Formula Weight 121.11
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Fluorobenzonitrile, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fre
EINECSNumber 214-784-9
RTECSNumber DI4368500
TSCA TSCA

RUO – Research Use Only