missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzonitrile, 99%

CAS: 1194-02-1 | C7H4FN | 121.114 g/mol

Supplier:  Thermo Scientific Chemicals A1302822

 View more versions of this product

Catalog No. AAA1302822


Only null left
Explore available promotions

Description

Description

4-Fluorobenzonitrile is used as chemical intermediate, solvent for perfumes and pharmaceuticals, stabilizer for chlorinated solvents, HPLC analysis, catalyst and component of transition-metal complex catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzonitrile is used as chemical intermediate, solvent for perfumes and pharmaceuticals, stabilizer for chlorinated solvents, HPLC analysis, catalyst and component of transition-metal complex catalysts.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4-Fluorobenzonitrile
34°C to 37°C
65°C (149°F)
C7H4FN
100 g
p-fluorobenzonitrile, benzonitrile, 4-fluoro, 4-cyanofluorobenzene, p-cyanofluorobenzene, benzonitrile, p-fluoro, 4-fluoro-benzonitrile, 4-fluorobenzenecarbonitrile, 4-fluorobenzonitril, pfbn, 1-cyano-4-fluorobenzene
AEKVBBNGWBBYLL-UHFFFAOYSA-N
4-fluorobenzonitrile
14517
99%
1194-02-1
186°C to 188°C
Almond-like
MFCD00001812
2041517
Insoluble in water.
C1=CC(=CC=C1C#N)F
121.114
121.11

Chemical Identifiers

1194-02-1
121.114
AEKVBBNGWBBYLL-UHFFFAOYSA-N
14517
C1=CC(=CC=C1C#N)F
C7H4FN
MFCD00001812
p-fluorobenzonitrile, benzonitrile, 4-fluoro, 4-cyanofluorobenzene, p-cyanofluorobenzene, benzonitrile, p-fluoro, 4-fluoro-benzonitrile, 4-fluorobenzenecarbonitrile, 4-fluorobenzonitril, pfbn, 1-cyano-4-fluorobenzene
4-fluorobenzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 214-784-9

RTECSNumber : DI4368500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only