missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzonitrile, 99%

Catalog No. AAA1302806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1302806 5 g
AAA1302814 25 g
AAA1302822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1302806 Supplier Thermo Scientific Chemicals Supplier No. A1302806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1194-02-1 | C7H4FN | 121.114 g/mol

4-Fluorobenzonitrile is used as chemical intermediate, solvent for perfumes and pharmaceuticals, stabilizer for chlorinated solvents, HPLC analysis, catalyst and component of transition-metal complex catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzonitrile is used as chemical intermediate, solvent for perfumes and pharmaceuticals, stabilizer for chlorinated solvents, HPLC analysis, catalyst and component of transition-metal complex catalysts.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1194-02-1
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.114
MDL Number MFCD00001812
InChI Key AEKVBBNGWBBYLL-UHFFFAOYSA-N
Synonym p-fluorobenzonitrile, benzonitrile, 4-fluoro, 4-cyanofluorobenzene, p-cyanofluorobenzene, benzonitrile, p-fluoro, 4-fluoro-benzonitrile, 4-fluorobenzenecarbonitrile, 4-fluorobenzonitril, pfbn, 1-cyano-4-fluorobenzene
PubChem CID 14517
IUPAC Name 4-fluorobenzonitrile
SMILES C1=CC(=CC=C1C#N)F

Specifications

Melting Point 34°C to 37°C
Boiling Point 186°C to 188°C
Flash Point 65°C (149°F)
Odor Almond-like
Quantity 5 g
Beilstein 2041517
Solubility Information Insoluble in water.
Formula Weight 121.11
Percent Purity 99%
Chemical Name or Material 4-Fluorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 214-784-9
RTECSNumber DI4368500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only