missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoic acid, 98%

Catalog No. AAA1412936
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1412936 500 g
AAA1412914 25 g
AAA1412922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1412936 Supplier Thermo Scientific Chemicals Supplier No. A1412936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 456-22-4 | C7H5FO2 | 140.113 g/mol

4-Fluorobenzoic acid is used as an intermediate for agrochemicals and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzoic acid is used as an intermediate for agrochemicals and pharmaceuticals.

Solubility
Soluble in alcohol, hot water, methanol, and ether. Very slightly soluble in cold water.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.

Chemical Identifiers

CAS 456-22-4
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.113
MDL Number MFCD00002530
InChI Key BBYDXOIZLAWGSL-UHFFFAOYSA-N
Synonym p-fluorobenzoic acid, benzoic acid, 4-fluoro, para-fluorobenzoic acid, 4-fluoro-benzoic acid, benzoic acid, p-fluoro, unii-v5roo2hou4, p-fluoro-benzoic acid, v5roo2hou4, 4-fluorobenzoatd, p-flurobenzoic acid
PubChem CID 9973
ChEBI CHEBI:20364
IUPAC Name 4-fluorobenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)F

Specifications

Melting Point 183°C to 187°C
Density 1.47
Quantity 500 g
Beilstein 1906922
Merck Index 14,4171
Solubility Information Soluble in alcohol,hot water,methanol,and ether. Very slightly soluble in cold water.
Formula Weight 140.11
Percent Purity 98%
Chemical Name or Material 4-Fluorobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-259-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only